• Início
  • Resultados da busca
  • Prebiotic effects of diet supplemented with the cultivated red seaweed Chondrus crispus or with fructo-oligo-saccharide on host immunity, colonic microbiota and gut microbial metabolites.
Prebiotic effects of diet supplemented with the cultivated red seaweed Chondrus crispus or with fructo-oligo-saccharide on host immunity, colonic microbiota and gut microbial metabolites.
BMC complementary and alternative medicine (2015-08-15)
Jinghua Liu, Saveetha Kandasamy, Junzeng Zhang, Christopher W Kirby, Tobias Karakach, Jeff Hafting, Alan T Critchley, Franklin Evans, Balakrishnan Prithiviraj
RESUMO
Gastrointestinal microbial communities are diverse and are composed of both beneficial and pathogenic groups. Prebiotics, such as digestion-resistant fibers, influence the composition of gut microbiota, and can contribute to the improvement of host health. The red seaweed Chondrus crispus is rich in dietary fiber and oligosaccharides, however its prebiotic potential has not been studied to date. Prebiotic effects were investigated with weaning rats fed a cultivated C. crispus-supplemented diet. Comparison standards included a fructo-oligo-saccharide (FOS) diet and a basal diet. The colonic microbiome was profiled with a 16S rRNA sequencing-based Phylochip array. Concentrations of short chain fatty acids (SCFAs) in the feacal samples were determined by gas chromatography with a flame ionization detector (GC-FID) analysis. Immunoglobulin levels in the blood plasma were analyzed with an enzyme-linked immunosorbent assay (ELISA). Histo-morphological parameters of the proximal colon tissue were characterized by hematoxylin and eosin (H&E) staining. Phylochip array analysis indicated differing microbiome composition among the diet-supplemented and the control groups, with the C. crispus group (2.5% supplementation) showing larger separation from the control than other treatment groups. In the 2.5% C. crispus group, the population of beneficial bacteria such as Bifidobacterium breve increased (4.9-fold, p=0.001), and the abundance of pathogenic species such as Clostridium septicum and Streptococcus pneumonia decreased. Higher concentrations of short chain fatty acids (i.e., gut microbial metabolites), including acetic, propionic and butyric acids, were found in faecal samples of the C. crispus-fed rats. Furthermore, both C. crispus and FOS supplemented rats showed significant improvements in proximal colon histo-morphology. Higher faecal moisture was noted in the 2.5% C. crispus group, and elevated plasma immunoglobulin (IgA and IgG) levels were observed in the 0.5% C. crispus group, as compared to the basal feed group. The results suggest multiple prebiotic effects, such as influencing the composition of gut microbial communities, improvement of gut health and immune modulation in rats supplemented with cultivated C. crispus.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
2-Ethylbutyric acid, 99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
2-Ethylbutyric acid, ≥98%, FCC, FG
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl