• Início
  • Resultados da busca
  • Additive effects of Artemisia capillaris extract and scopoletin on the relaxation of penile corpus cavernosum smooth muscle.
Additive effects of Artemisia capillaris extract and scopoletin on the relaxation of penile corpus cavernosum smooth muscle.
International journal of impotence research (2015-10-09)
B R Choi, S K Kumar, C Zhao, L T Zhang, C Y Kim, S W Lee, J-H Jeon, K K Soní, I So, S H Kim, N C Park, H K Kim, J K Park
RESUMO
The objective was to investigate the cellular effect and action mechanism of Artemisia capillaris extract (ACE) and its component, scopoletin, on penile corpus cavernosum smooth muscle (PCCSM). In vitro study with PCCSM, the precontracted PCCSM with phenylephrine was treated with ACE or scopoletin. Cyclic nucleotides in the perfusate were measured by radioimmunoassay and expression of protein and mRNA of endothelial nitric oxide synthase (eNOS) and neuronal nitric oxide synthase in the perfused PCCSM were measured by western blot and real-time PCR, respectively. The interaction of ACE or scopoletin with udenafil was also evaluated. ACE and scopoletin exerted a significant and concentration-dependent relaxation in PCCSM. The perfusion with ACE or scopoletin significantly increased cyclic adenosine monophosphate (cAMP) and cyclic guanosine monophosphate (cGMP) and the perfusion with ACE or scopoletin increased the expression of eNOS mRNA and protein. Furthermore, ACE or scopoletin enhanced udenafil-inducing relaxation in PCCSM. ACE and scopoletin relaxed the PCCSM mainly by activating nitric oxide-cGMP system and cAMP pathway and they may be additive therapeutic candidates for ED patients who do not completely respond to udenafil.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Chloromethane solution, 200 μg/mL in methanol, analytical standard
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent