• Início
  • Resultados da busca
  • Identification of Small Aliphatic Aldehydes in Pretreated Lignocellulosic Feedstocks and Evaluation of Their Inhibitory Effects on Yeast.
Identification of Small Aliphatic Aldehydes in Pretreated Lignocellulosic Feedstocks and Evaluation of Their Inhibitory Effects on Yeast.
Journal of agricultural and food chemistry (2015-11-04)
Adnan Cavka, Stefan Stagge, Leif J Jönsson
RESUMO
Six lignocellulosic hydrolysates produced through acid pretreatment were analyzed for the occurrence of formaldehyde, acetaldehyde, and glycolaldehyde. Acetaldehyde was found in all six (0.3-1.6 mM) and formaldehyde in four (≤ 4.4 mM), whereas glycolaldehyde was not detected. To assess the relevance of these findings, fermentations with yeast and formaldehyde or acetaldehyde were performed in the concentration interval 0.5-10 mM. Formaldehyde already inhibited at 1.0 mM, whereas 5.0 mM acetaldehyde was needed to obtain a clear inhibitory effect. After 24 h of fermentation, 1.5 mM formaldehyde reduced the glucose consumption by 85%, the balanced ethanol yield by 92%, and the volumetric productivity by 91%. The results show that formaldehyde and acetaldehyde are prevalent in pretreated lignocellulose and that formaldehyde in some cases could explain a large part of the inhibitory effects on yeast by lignocellulosic hydrolysates, as three of six hydrolysates contained ≥ 1.9 mM formaldehyde, which was shown to be strongly inhibitory.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Purine, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required