  • Search Results
  • YGS40, an active fraction of Yi-Gan San, reduces hydrogen peroxide-induced apoptosis in PC12 cells.
YGS40, an active fraction of Yi-Gan San, reduces hydrogen peroxide-induced apoptosis in PC12 cells.
Chinese journal of natural medicines (2015-06-16)
Yu-Rong Zhao, Wei Qu, Wen-Yuan Liu, Hao Hong, Feng Feng, Han Chen, Ning Xie
ABSTRACT
In our previous study, we have elucidated the chemical profile of YGS40, a fraction of Yi-Gan San (YGS), used for the treatment of Alzheimer's disease (AD). Oxidative stress-induced apoptosis is implicated in neurodegenerative disorders such as AD. The aim of the present study was to explore the protective effects of YGS40 against hydrogen peroxide (H2O2)-induced apoptosis in PC12 cells and the underlying mechanisms. PC12 cells were exposed to 100 μmol·L(-1) of H2O2 for 12 h with or without YGS40 pretreatment. Cytotoxicity was determined by MTT (3, (4, 5-dimethylthiazole-2-yl) 2, 5-diphenyl-tetrazolium bromide) and lactate dehydrogenase (LDH) release assays; apoptosis was detected by Annexin V/propidium iodide coupled staining and by determining caspase-3 activity and Bax and Bcl-2 protein levels. Mitochondrial membrane potential (MMP) was assessed by the retention of rhodamine123; and the activities of superoxide dismutase (SOD) and malondialdehyde (MDA) were measured using commercially available enzymatic kits. Pretreatment with YGS40 significantly prevented H2O2-induced cytotoxicity and protected the cells against H2O2-triggered apoptosis characterized by externalization of membrane phosphatidylserine and caspase-3 activation and the increased ratio of Bax/Bcl-2 in PC12 cells. Further studies showed that YGS40 suppressed H2O2-induced MMP loss, increased SOD activity, and decreased MDA level. These findings suggest that YGS40 may be beneficial for the prevention and treatment of oxidative stress-mediated disorders.
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, natural, 97%, FG