  • Search Results
  • Increased Heterologous Protein Expression in Drosophila S2 Cells for Massive Production of Immune Ligands/Receptors and Structural Analysis of Human HVEM.
Increased Heterologous Protein Expression in Drosophila S2 Cells for Massive Production of Immune Ligands/Receptors and Structural Analysis of Human HVEM.
Molecular biotechnology (2015-07-24)
Weifeng Liu, Vladimir Vigdorovich, Chenyang Zhan, Yury Patskovsky, Jeffrey B Bonanno, Stanley G Nathenson, Steven C Almo
ABSTRACT
Many immune ligands and receptors are potential drug targets, which delicately manipulate a wide range of immune responses. We describe here the successful application of an efficient method to dramatically improve the heterologous expression levels in Drosophila Schneider 2 cells, which enables the high-throughput production of several important immune ligands/receptors for raising antibodies, and for the structural and functional analyses. As an example, we purified the protein and characterized the structure of the immune receptor herpesvirus entry mediator (HVEM, TNFRSF14). HVEM is a member of tumor necrosis factor receptor superfamily, which is recognized by herpes simplex virus glycoprotein D (gD) and facilitates viral entry. HVEM participates in a range of interactions with other cell surface molecules, including LIGHT, BTLA, and CD160 to modulate a wide range of immune processes in CD4(+) and CD8(+) T cells, as well as NK cells. Due to the involvement of HVEM in these diverse signaling interactions, crystal structures of HVEM in complex with gD or BTLA have been previously reported. Here, we report the structure of HVEM in the absence of any ligands.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
SAFC
Tromethamine
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Sigma-Aldrich
Methanol, NMR reference standard
SAFC
Glycine
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%