  • Search Results
  • Placental perfusion in uterine ischemia model as evaluated by dynamic contrast enhanced MRI.
Placental perfusion in uterine ischemia model as evaluated by dynamic contrast enhanced MRI.
Journal of magnetic resonance imaging : JMRI (2015-04-10)
Alexander Drobyshevsky, P V Prasad
ABSTRACT
To validate DCE MRI method of placental perfusion estimation and to demonstrate application of the method in a rabbit model of fetal antenatal hypoxia-ischemia. Placental perfusion was estimated by dynamic contrast imaging with bolus injection of Gd-DTPA in 3 Tesla GE magnet in a rabbit model of placental ischemia-reperfusion in rabbit dams at embryonic day 25 gestation age. Placental perfusion was measured using steepest slope method on DCE MRI before and after intermittent 40 min uterine ischemia. Antioxidants (n = 2 dams, 9 placentas imaged) or vehicle (n = 5 dams, 23 placenta imaged) were given systemically in a separate group of dams during reperfusion-reoxygenation. Placental perfusion was also measured in two dams from the antioxidant group (10 placentas) and two dams from the control group (12 placentas) by fluorescent microspheres method. While placental perfusion estimates between fluorescent microspheres and DCE MRI were significantly correlated (R(2)  = 0.85; P < 0.01), there was approximately 33% systematic underestimation by the latter technique. DCE MRI showed a significant decrease in maternal placental perfusion in reperfusion-reoxygenation phase in the saline, 0.44 ± 0.06 mL/min/g (P = 0.012, t-test), but not in the antioxidant group, 0.62 ± 0.06 mL/min/g, relative to pre-occlusion values (0.77 ± 0.07 and 0.84 ± 0.12 mL/min/g, correspondingly). Underestimation of true perfusion in placenta by steepest slope DCE MRI is significant and the error appears to be systematic.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
2-Ethoxyethyl acetate, 98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications