Merck
Identification of invasion proteins of Cryptosporidium parvum.
World journal of microbiology & biotechnology (2015-10-24)
Preeti Singh, Bijay Ranjan Mirdha, Alagiri Srinivasan, Lokesh A Rukmangadachar, Sundeep Singh, Pratibha Sharma, Gururao Hariprasad, Hariprasad Gururao, Hariprasad Gururao, Kalpana Luthra
ABSTRACT
Host cell interactions and invasion by Cryptosporidium is a complex process mediated by zoites ligand-host cell receptors. Knowledge of proteins involved in this process will enable entry level inhibitors to be tried as therapeutic agents. In the present study, invasion proteins of Cryptosporidium parvum were studied in vitro. Cryptosporidium sporozoites membrane proteins were isolated and Cy5 dye labelled. They were then allowed to interact with the intact host cells. The interacting proteins were identified using 2-dimensional gel electrophoresis followed by mass spectrometry analysis. Sixty-one proteins were identified including twenty-seven previously reported invasion proteins. The newly identified proteins such as serine/threonine protein kinase, PI4 kinase, Hsp105 and coiled coil may have their roles in the parasitic invasion process. Thus, a new approach was used in the study to identify the probable proteins involved in invasion and/or host-parasite interactions. The advantage of this method is that it takes only a months' time instead of decades to identify these proteins involved in invasion process.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)