  • Search Results
  • Melatonin attenuates impairments of structural hippocampal neuroplasticity in OXYS rats during active progression of Alzheimer's disease-like pathology.
Melatonin attenuates impairments of structural hippocampal neuroplasticity in OXYS rats during active progression of Alzheimer's disease-like pathology.
Journal of pineal research (2015-05-20)
Natalia A Stefanova, Kseniya Y Maksimova, Elena Kiseleva, Ekaterina A Rudnitskaya, Natalia A Muraleva, Nataliya G Kolosova
ABSTRACT
Translational research on Alzheimer's disease (AD) has often focused on reducing the high cerebral levels of amyloid-β (Aβ) as a key characteristic of AD pathogenesis. There is, however, a growing body of evidence that synaptic dysfunction may be crucial for the development of the most common (sporadic) form of AD. The applicability of melatonin (mainly produced by the pineal gland) to the treatment of AD is actively evaluated, but usually, such studies are based on animal models of early-onset AD, which is responsible for only ~5% of AD cases. We have shown previously that in OXYS rats (an established model of sporadic AD), accumulation of toxic forms of Aβ in the brain occurs later than does the development of signs of neurodegenerative changes and synaptic failure. In this regard, recently, we uncovered beneficial neuroprotective effects of melatonin (prophylactic dietary supplementation) in OXYS rats. Our aim here was to evaluate, starting at the age of active progression of AD-like pathology in OXYS rats, the effects of long-term oral administration of melatonin on the structure of synapses and on neuronal and glial cells of the hippocampus. Melatonin significantly increased hippocampal synaptic density and the number of excitatory synapses, decreased the number of inhibitory synapses, and upregulated pre- and postsynaptic proteins (synapsin I and PSD-95, respectively). Furthermore, melatonin improved the ultrastructure of neuronal and glial cells and reduced glial density. Based on our past and present results, the repair of neuroplasticity by melatonin is a promising strategy against AD.
Product Description
SAFC
Tromethamine
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
SAFC
Glycine
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free