  • Search Results
  • Human placenta-derived multipotent mesenchymal stromal cells involved in placental angiogenesis via the PDGF-BB and STAT3 pathways.
Human placenta-derived multipotent mesenchymal stromal cells involved in placental angiogenesis via the PDGF-BB and STAT3 pathways.
Biology of reproduction (2015-09-12)
Cheng-Yi Chen, Shu-Hsiang Liu, Chia-Yu Chen, Pei-Chun Chen, Chie-Pein Chen
ABSTRACT
We studied the smooth muscle cell differentiation capability of human placental multipotent mesenchymal stromal cells (hPMSCs) and identified how endothelial cells recruit hPMSCs participating in vessel formation. hPMSCs from term placentas were induced to differentiate into smooth muscle cells under induction conditions and different matrix substrates. We assessed endothelial cells from umbilical veins for platelet-derived growth factor (PDGF)-BB expression and to induce hPMSC PDGFR-beta and STAT3 activation. Endothelial cells were co-cultured with hPMSCs for in vitro angiogenesis. Cell differentiation ability was then further assessed by mouse placenta transplantation assay. hPMSCs can differentiate into smooth muscle cells; collagen type I and IV or laminin support this differentiation. Endothelial cells expressed significant levels of PDGF-BB and activated STAT3 transcriptional activity in hPMSCs. Endothelial cell-conditioned medium induced hPMSC migration, which was inhibited by STAT3 small interfering RNA transfection or by pretreatement with PDGFR-beta-blocking antibody but not by PDGFR-alpha-blocking antibody or isotype immunoglobulin G (IgG; P < 0.001). hPMSCs can incorporate into endothelial cells with tube formation and promote endothelial cells, forming capillary-like networks than endothelial cells alone (tube lengths: 12 024.1 ± 960.1 vs. 9404.2 ± 584.7 pixels; P < 0.001). Capillary-like networks were significantly reduced by hPMSCs pretreated with PDGFR-beta-blocking antibody but not by PDGFR-alpha-blocking antibody or isotype IgG (P < 0.001). Transplantation of hPMSCs into mouse placentas revealed incorporation of the hPMSCs into vessel walls, which expressed alpha-smooth muscle actin, calponin, and smooth muscle myosin (heavy chain) in vivo. In conclusion, endothelial cell-hPMSC interactions occur during vessel development of placenta. Placental endothelial cell-derived PDGF-BB recruits hPMSCs involved in vascular development via PDGFR-beta/STAT3 activation.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Calcium phosphate, purum p.a., ≥96.0% (calc. as Ca3(PO4)2, KT)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications