  • Search Results
  • Antioxidant Properties of Aqueous Extract of Roasted Hulled Barley in Bulk Oil or Oil-in-Water Emulsion Matrix.
Antioxidant Properties of Aqueous Extract of Roasted Hulled Barley in Bulk Oil or Oil-in-Water Emulsion Matrix.
Journal of food science (2015-09-27)
Sumi Oh, Mi-Ja Kim, Kye Won Park, Jae Hwan Lee
ABSTRACT
Antioxidant properties of the aqueous extracts of hulled barley (Hordeum vulgare L.) that had been roasted at 210 °C for 20 min were determined in bulk oil and oil-in-water (O/W) emulsions. Bulk oils were heated at 60, 100, and 180 °C, and O/W emulsions were oxidized under riboflavin photosensitization. The content of phenolic compounds was analyzed by high-performance liquid chromatography, and in vitro antioxidant assays were also conducted. The major phenolics contained in the aqueous extract of roasted hulled barley (AERB) were p-coumaric, ferulic, protocatechuic, chlorogenic, 4-hydroxybenzoic, and vanillic acids. Depending on the concentration and oxidation temperature, AERB had antioxidant or prooxidant properties in bulk oil. At 60 °C, AERB at a concentration of 0.5% acted as a prooxidant, whereas at 1.0% it acted as an antioxidant. At 100 °C, AERB acted as an antioxidant irrespective of concentration. In 180 °C conditions, 0.5% AERB acted as a prooxidant, whereas other concentrations of AERB acted as antioxidants. In the case of riboflavin photosensitized O/W emulsions, AERB showed antioxidant properties irrespective of concentration. Antioxidant abilities of AERB are affected by the food matrix, including bulk oil and O/W emulsions, and concentrations of AERB, even though diverse phenolic compounds may display high antioxidant properties in in vitro assays. Roasted barley has been widely used as a tea ingredient in East Asian countries such as Korea, China, and Japan. The highly antioxidative properties of the aqueous extracts of roasted barley have been confirmed in bulk oil and O/W emulsions as well as in vitro assays because of the presence of phenolic compounds. The results of this study can contribute to the development of antioxidant-rich beverages using roasted barley by aiding in the selection of proper food matrix-containing extracts of high phenolic compounds, as well as by expanding consumers’ choices for healthy beverages.
Sigma-Aldrich
p-Anisidine, 99%
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Tannic acid, ACS reagent
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Tannic acid, Source: Chinese natural gall nuts
Sigma-Aldrich
Tannic acid
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Tannic acid, puriss., meets analytical specification of USP, powder
Sigma-Aldrich
Aluminum chloride, anhydrous, sublimed, ≥98%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Potassium acetate, meets USP testing specifications
Sigma-Aldrich
Potassium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium acetate, 98%
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)