  • Search Results
  • MicroRNA-204 Targets Runx2 to Attenuate BMP-2-induced Osteoblast Differentiation of Human Aortic Valve Interstitial Cells.
MicroRNA-204 Targets Runx2 to Attenuate BMP-2-induced Osteoblast Differentiation of Human Aortic Valve Interstitial Cells.
Journal of cardiovascular pharmacology (2015-03-26)
Yongjun Wang, Si Chen, Cheng Deng, Fei Li, Yin Wang, Xingjian Hu, Feng Shi, Nianguo Dong
ABSTRACT
Osteoblast differentiation of valve interstitial cells (VICs) is a key step in valve calcification, but the molecular mechanisms involved are not fully understood. In this study, we aimed to investigate whether microRNA (miR)-204-regulated VICs differentiation through modulation of runt-related transcription factor 2 (Runx2), a key transcription factor for osteogenesis. Our data demonstrated that miR-204 was markedly downregulated in both human calcified aortic valves and bone morphogenetic protein (BMP)-2-stimulated aortic VICs. In vitro experiments showed that miR-204 acted as a negative regulator of osteogenic differentiation by repressing Runx2 and thereby inhibiting expression of osteoblast-related genes, including alkaline phosphatase and osteocalcin, which were all induced by BMP-2. Luciferase reporter assays validated Runx2 as the direct target of miR-204. Furthermore, increased alkaline phosphatase activity and osteocalcin expression after miR-204 inhibition were abolished by small interfering RNA-mediated silencing of Runx2. Overall, these data suggested miR-204 as a possible molecular switch inhibiting osteoblastic transdifferentiation of human aortic VICs and targeting miR-204 may have therapeutic potential for human aortic valve calcification.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, BMP-2, recombinant, expressed in HEK 293 cells, HumanKine, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, Carrier Free, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
BMP-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
BMP-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)