  • Search Results
  • Rapid tumor induction in zebrafish by TALEN-mediated somatic inactivation of the retinoblastoma1 tumor suppressor rb1.
Rapid tumor induction in zebrafish by TALEN-mediated somatic inactivation of the retinoblastoma1 tumor suppressor rb1.
Scientific reports (2015-09-09)
Staci L Solin, Heather R Shive, Kevin D Woolard, Jeffrey J Essner, Maura McGrail
ABSTRACT
Investigating the in vivo role of tumor suppressor genes in cancer is technically challenging due to their essential requirement during early animal development. To address this bottleneck, we generated genetic mosaic adult zebrafish using TALEN genome editing and demonstrate somatic inactivation of the tumor suppressor retinoblastoma1 (rb1) induces tumorigenesis at high frequency. 11-33% of 1-cell stage embryos injected with TALEN mRNAs targeting rb1 exon 2 or 3 develop tumors beginning as early as 3.5 months of age. Lesions predominantly arise in the brain and show features of neuroectodermal-like and glial-like tumors. Mutant allele analysis is consistent with tumor initiation due to somatic inactivation of rb1, revealing a conserved role for rb1 in tumor suppression across vertebrates. In contrast to genetic mosaics, heterozygous rb1-/+ adults show no evidence of neoplasia, while homozygous mutant rb1-/- are larval lethal. This is the first demonstration that somatic inactivation of a tumor suppressor causes cancer in zebrafish, and highlights the utility of site-specific nucleases to create genetic mosaic zebrafish for tumor suppressor gene discovery. Somatic inactivation with site-directed nucleases in zebrafish presents a rapid and scalable strategy to study tumor suppressor gene function in cancer.
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Eosin Y
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission