  • Search Results
  • Discovery of highly conserved unique peanut and tree nut peptides by LC-MS/MS for multi-allergen detection.
Discovery of highly conserved unique peanut and tree nut peptides by LC-MS/MS for multi-allergen detection.
Food chemistry (2015-10-17)
Jennifer Sealey-Voyksner, Jerry Zweigenbaum, Robert Voyksner
ABSTRACT
Proteins unique to peanuts and various tree nuts have been extracted, subjected to trypsin digestion and analysis by liquid chromatography/quadrupole time-of-flight mass spectrometry, in order to find highly conserved peptides that can be used as markers to detect peanuts and tree nuts in food. The marker peptide sequences chosen were those found to be present in both native (unroasted) and thermally processed (roasted) forms of peanuts and tree nuts. Each peptide was selected by assuring its presence in food that was processed or unprocessed, its abundance for sensitivity, sequence size, and uniqueness for peanut and each specific variety of tree nut. At least two peptides were selected to represent peanut, almond, pecan, cashew, walnut, hazelnut, pine nut, Brazil nut, macadamia nut, pistachio nut, chestnut and coconut; to determine the presence of trace levels of peanut and tree nuts in food by a novel multiplexed LC-MS method.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Ultrapure Acetonitrile
SAFC
Tromethamine
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)