  • Search Results
  • Vitamin B₂ Sensitizes Cancer Cells to Vitamin-C-Induced Cell Death via Modulation of Akt and Bad Phosphorylation.
Vitamin B₂ Sensitizes Cancer Cells to Vitamin-C-Induced Cell Death via Modulation of Akt and Bad Phosphorylation.
Journal of agricultural and food chemistry (2015-07-15)
Ni Chen, Shutao Yin, Xinhua Song, Lihong Fan, Hongbo Hu
ABSTRACT
Vitamin C is an essential dietary nutrient that has a variety of biological functions. Recent studies have provided promising evidence for its additional health benefits, including anticancer activity. Vitamin B2, another essential dietary nutrient, often coexists with vitamin C in some fruits, vegetables, or dietary supplements. The objective of the present study is to determine whether the combination of vitamin C and B2 can achieve a synergistic anticancer activity. MDA-MB-231, MCF-7, and A549 cells were employed to evaluate the combinatory effects of vitamin C and B2. We found that the combination of vitamin C and B2 resulted in a synergistic cell death induction in all cell lines tested. Further mechanistic investigations revealed that vitamin B2 sensitized cancer cells to vitamin C through inhibition of Akt and Bad phosphorylation. Our findings identified vitamin B2 as a promising sensitizer for improving the efficacy of vitamin-C-based cancer chemoprevention and chemotherapy.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
3-Ethoxy-5,6-dibromosalicylaldehyde, ≥95% (HPLC)