  • Search Results
  • Simultaneous determination of four phenolic acids and seven alkaloids in rat plasma after oral administration of traditional Chinese medicinal preparation Jinqi Jiangtang Tablet by LC-ESI-MS/MS.
Simultaneous determination of four phenolic acids and seven alkaloids in rat plasma after oral administration of traditional Chinese medicinal preparation Jinqi Jiangtang Tablet by LC-ESI-MS/MS.
Journal of pharmaceutical and biomedical analysis (2015-09-05)
Yan-xu Chang, Ai-hua Ge, Xie-an Yu, Xiu-cheng Jiao, Jin Li, Jun He, Ji Tian, Wei Liu, John Teye Azietaku, Bo-li Zhang, Xiu-mei Gao
ABSTRACT
A rapid, sensitive and selective high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for the simultaneous determination of four phenolic acids (neochlorogenic acid, chlorogenic acid, cryptochlorogenic acid and ferulic acid) and seven alkaloids (berberine, epiberberine, coptisine, magnoflorine, berberubine, palmatine and jatrorrhizine) in rat plasma. After mixing with the internal standards tetrahydropalmatine (IS1) and rosmarinic acid (IS2), plasma samples were pretreated by protein precipitation using acetonitrile. The HPLC analysis was performed on an Agilent Eclipse plus C18 (4.6 mm×100 mm, 1.8 μm) column with mobile phase consisting of 0.1% formic acid aqueous solution and acetonitrile at a flow rate of 0.3 mL min(-1). The detection was accomplished for the analytes and internal standards using positive electrospray ionization for the alkaloids and negative electrospray ionization for the phenolic acids in multiple-reaction monitoring mode. The method showed a good linearity over a wide concentration range (r(2)>0.99). The lower limit of quantification of seven alkaloids was lower than 2 ng mL(-1) and that of four phenolic acids was less than 20 ng mL(-1). The developed method was applied to the pharmacokinetic study of 11 components after oral administration of traditional Chinese medicinal preparation Jinqi Jiangtang Tablet in rats.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)