• Início
  • Resultados da busca
  • Probing cell-matrix interactions in RGD-decorated macroporous poly (ethylene glycol) hydrogels for 3D chondrocyte culture.
Probing cell-matrix interactions in RGD-decorated macroporous poly (ethylene glycol) hydrogels for 3D chondrocyte culture.
Biomedical materials (Bristol, England) (2015-06-25)
Jingjing Zhang, Ayeesha Mujeeb, Yanan Du, Jianhao Lin, Zigang Ge
RESUMO
Macroporous hydrogels have shown great promise as scaffolds for cartilage engineering by facilitating nutrition transport and tissue in growth. Cell-matrix adhesion-a fundamental process in tissue engineering-has shown a profound effect on subsequent cell phenotype, extracellular matrix (ECM) accumulation, and tissue reorganization. In this study, arginine-glycine-aspartic acid (RGD) was introduced to macroporous hydrogels of poly (ethylene glycol) (PEG) to fabricate PEG-G400 (with 0.4mM RGD) and PEG-G2000 (2mM RGD) to probe the cell-matrix interactions within hydrogels. Primary chondrocytes demonstrated a slightly stretched morphology with increasing RGD concentration and PEG-G2000 hydrogels boosted cell viability, proliferation, and deposition of collagen II and GAG, in comparison to the PEG-G400 and PEG-RED groups. Results also revealed chondrocytes within the cell aggregates underwent dedifferentiation and hypertrophy within RGD incorporated hydrogels, as evidenced by the high level of gene expression of collagen I on day 14 and strong immunohistological staining of collagen X and collagen I on day 35. Evidently, a high concentration of RGD (2mM RGD) enhanced cell-matrix interactions through elevating the expression of integrin β1 and vinculin. Thus, the integration of RGD in macroporous hydrogels with a concentration of 2 mM may be sufficient for improving cell functionality, with a slight probability of dedifferentiation and hypertrophy of chondrocytes.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ~99%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
1,9-Dimethyl-Methylene Blue zinc chloride double salt, Dye content 80 %
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%