  • Search Results
  • CD80-CD28 signaling controls the progression of inflammatory colorectal carcinogenesis.
CD80-CD28 signaling controls the progression of inflammatory colorectal carcinogenesis.
Oncotarget (2015-01-18)
Marco Scarpa, Paola Brun, Melania Scarpa, Susan Morgan, Andrea Porzionato, Andromachi Kotsafti, Marina Bortolami, Andrea Buda, Renata D'Incà, Veronica Macchi, Giacomo C Sturniolo, Massimo Rugge, Romeo Bardini, Ignazio Castagliuolo, Imerio Angriman, Carlo Castoro
ABSTRACT
In patients with ulcerative colitis (UC) the cumulative risk of colon cancer is lower than the actual rate of dysplasia suggesting an efficient immune surveillance mechanism. Since the co-stimulatory molecule CD80 is overexpressed in dysplastic colonic mucosa of UC patients and T-cell activation entails effective costimulation, we aimed to evaluate the functional implication of CD80 signaling in colonic UC-associated carcinogenesis. In humans, we observed that the percentage of CD80+ and HLA-A+ IEC was increased in the dysplastic colonic mucosa of UC patients. In vitro, IEC activated CD8+ T-cells through a CD80-dependent pathway. Finally, in the AOM/DSS-induced colonic adenocarcinoma model CD80 signaling inhibition significantly increased the frequency and extension of high-grade dysplasia, whereas enhancing CD80 activity with an anti-CTLA4 antibody significantly decreased colonic dysplasia. In conclusion, CD80 signaling between IEC and T-cells represents a key factor controlling the progression from low to high grade dysplasia in inflammatory colonic carcinogenesis.
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Azoxymethane, 13.4 M, ≥98%
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture