  • Search Results
  • The broad pattern recognition spectrum of the Toll-like receptor in mollusk Zhikong scallop Chlamys farreri.
The broad pattern recognition spectrum of the Toll-like receptor in mollusk Zhikong scallop Chlamys farreri.
Developmental and comparative immunology (2015-06-01)
Mengqiang Wang, Lingling Wang, Ying Guo, Rui Sun, Feng Yue, Qilin Yi, Linsheng Song
ABSTRACT
Toll-like receptors (TLRs) are among the most studied pattern recognition receptors (PRRs) playing essential roles in innate immune defenses. In the present study, the basic features of CfTLR in mollusk Zhikong scallop Chlamys farreri, including sequence homology, tissue distribution, subcellular localization and ligands spectrum, were investigated to elucidate its pattern recognition. The elements of extracellular domains (ECD) in CfTLR displayed high homology to the corresponding parts of the ECDs in TLRs from Homo sapiens. CfTLR protein was detected in hemocytes, mantle, gills, hepatopancreas, kidney and gonad of the scallops, and it was localized in both the plasma membranes and the lysosomes in HEK293T cells. CfTLR could activate NFκB in response to multiple HsTLR ligands including Pam3CSK4, glucan (GLU), peptidoglycan (PGN), polyriboinosinic:polyribocytidylic acid (poly I:C), Imiquimod and three types of CpG. Additionally, the scallop serum could enhance the induction of NFκB in the CfTLR expressing cells elicited by most PAMPs, including GLU, PGN, Imiquimod and four types of CpG. It could be concluded that this primitive mollusk TLR shared a hybrid function in pattern recognition and could recognize broader ligands than mammalian TLRs, and its mosaic capability of pathogen associated molecular pattern (PAMP) recognition might be based on the basic features of its structure, ligand properties and the assistance of some components in scallop serum.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Peptidoglycan from Staphylococcus aureus, cell wall component
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Imiquimod, ≥98% (HPLC), solid
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Polyinosinic–polycytidylic acid sodium salt, TLR ligand tested
SAFC
Isopropyl β-D-1-thiogalactopyranoside