BR
PT
Aplicações
Produtos
Serviços
Suporte
Home
Search Results
Highly efficient quantum dot near-infrared light-emitting diodes.
Highly efficient quantum dot near-infrared light-emitting diodes.
Nature Photonics (2016)
Gong X, et al.