Lutein exerts anti-inflammatory effects in patients with coronary artery disease.
Atherosclerosis (2017-05-21)
Rosanna W S Chung, Per Leanderson, Anna K Lundberg, Lena Jonasson
RESUMO
Many coronary artery disease (CAD) patients exhibit chronic low-grade inflammation. Carotenoids are anti-oxidants with potential anti-inflammatory properties. Here, we first assessed relationships between interleukin (IL)-6 and individual carotenoids in plasma from CAD patients. Based on the results, we proceeded to assess anti-inflammatory effects of one carotenoid, lutein, in peripheral blood mononuclear cells (PBMCs) from CAD patients. Lutein + zeaxanthin (isomers with lutein being dominant), β-cryptoxanthin, lycopene, α- and β-carotene and IL-6 were measured in plasma from 134 patients with stable angina (SA) and 59 patients with acute coronary syndrome. In 42 patients, plasma measurements were also performed 3 months after coronary intervention. PBMCs from SA patients were pre-treated with lutein (1, 5 and 25 μM) for 24 h followed by 24 h incubation ± lipopolysaccharide (LPS). Cell pellets were collected for IL-6, IL-1β and TNF mRNA and intracellular lutein. Cytokine secretion was measured in cell media. Only lutein + zeaxanthin were inversely correlated with IL-6 in SA patients at baseline (r = -0.366, p < 0.001) and follow-up (r = -0.546, p < 0.001). Ex vivo, lutein was taken up by PBMCs from SA patients in a dose- and time-dependent manner. Pre-treatment with lutein dose-dependently lowered LPS-induced secretion of IL-6, IL-1β (p < 0.01) and TNF (p < 0.05), and also reduced IL-6, IL-1β and TNF mRNA expression (p < 0.05). Clinical findings highlighted the inverse association between lutein and IL-6 in CAD patients. Anti-inflammatory effects of lutein in PBMCs from CAD patients were consolidated in ex vivo experiments. Taken together, these results show that lutein has the potential to play a role in resolution of chronic inflammation in CAD patients.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Lutein, analytical standard