Merck
Monomers
Illustration of monomer
Monomers serve as the essential building blocks of polymers, influencing their properties, including strength, flexibility, and chemical resistance. They play a vital role in formulation of coatings, adhesives, and ophthalmic products like contact lenses (silane and hydrophilic monomers). Additionally, monomers are essential in dental restoration materials, plastics, elastomers, biodegradable polymers, and various industrial applications.
  • Monomers in Dental Materials, Contact Lenses, Wound Healing, and Cosmetic Fillers
  • Monomers for Polymer Production
  • Monomers for Coatings and Adhesives
  • Monomers for Smart Polymers
  • Bio-Based Monomers
Products
Sort by Relevance
Showing 1-20 of 533
Change View
Showing 1-20 of 533
Compare
Product Number
Product Name
Product Description
Pricing
Monomers in Dental Materials, Contact Lenses, Wound Healing, and Cosmetic Fillers
Monomers used in dental applications are biocompatible, wear-resistant and durable, making them ideal for dental adhesives and composites in restorative procedures. Ethylene oxide is a key monomer in the production of polyethylene glycol (PEG) and polyethylene oxide, which are used in medical devices, dental implants, biosensors, diagnostic applications, ophthalmic contact lenses, and optical coatings. 
Acrylates and methacrylate monomers such as 1,1,1-trimethylolpropane triacrylate, 1,1,1-trimethylolpropane trimethacrylate, 1,1,1-trimethylolethane trimethacrylate, ethylene glycol dimethacrylate, and triethylene glycol dimethacrylate, are widely used in dental applications for their adhesive and polymerization properties.
Monomers for Contact Lenses
Ethylene glycol dimethacrylate and 1-vinyl-2-pyrrolidone are utilized to facilitate the sustained release of timolol and hyaluronic acid from contact lenses for glaucoma treatment. 2-Ethylhexyl acrylate functions as a tackiness modifier in contact lens formulation, enhancing comfort and performance.
Monomers for Wound Healing and Dermal Fillers
1,4-butanediol diglycidyl ether serves as a crosslinking agent in wound healing applications, especially in hyaluronic acid (HA)-based dressings and gels, as well as in the preparation of heparosan crosslinked hydrogels, which are used in injectable dermal fillers.
Hydrogels
Hydrogels are formed from hydrophilic monomers and/or hydrophobic monomers. Hydrophilic monomers are essential for creating the water-absorbing characteristics of hydrogels, while hydrophobic monomers can enhance structural integrity and control swelling behavior. The hydrophilic monomers can retain moisture and create a biocompatible environment. This makes them ideal for contact lenses, as well as ophthalmic devices, drug delivery systems, wound dressings, and tissue engineering scaffolds.
Monomers for Polymer Production
Monomers are the fundamental building blocks of polymers, which are large molecules composed of repeating units. The choice of monomers determines key polymer properties such as strength, flexibility, thermal stability, and chemical resistance. 
Monomers for Coatings and Adhesives
Monomers are essential in formulating coatings and adhesives with tailored performance characteristics. Acrylic monomers are used in water-based paints and coatings, offering durability and UV-resistance. Epoxy monomers provide strong bonding and chemical resistance, making them ideal for adhesives and coatings in the construction and automotive industries. Additionally, UV absorbing monomers are essential in ophthalmic applications, enhancing protection and performance.
Coatings on Ophthalmic Devices
UV blockers in the ophthalmic industry are specialized materials or coatings designed to absorb or reflect ultraviolet (UV) radiation, protecting the eyes from harmful UV exposure. These blockers are essential in various ophthalmic products, including eyeglasses, contact lenses, and intraocular lenses. One notable example is the monomer 2-(4-Benzoyl-3-hydroxyphenoxy)ethyl acrylate, which is used as UV-blocking coating for contact lenses.
Monomers in Synthesis of Nanomaterials
Monomers play a crucial role in synthesizing nanomaterials, including nanoparticles with unique properties. These nanoparticles are widely used in drug delivery, medical imaging, and sensing applications, offering enhanced functionality at the nanoscale.
Monomers for Smart Polymers
Monomers can be polymerized to create smart polymers, which exhibit stimuli-responsive properties. These monomers are designed to react to external stimuli such as temperature, pH, light, or electric fields.
One widely used monomer for thermo-responsive polymers is N-Isopropylacrylamide (NIPAM). It exhibits a lower critical solution temperature (LCST) and transitions from hydrophilic to hydrophobic, precipitating out of solution when heated above a certain temperature. Similarly, acrylic acid monomers are polymerized to produce pH-sensitive polymers, which swell or shrink based on the acidity or basicity of their environment.
Thermo-Responsive Polymers
Poly(N-isopropylacrylamide) (PNIPAM) is a thermo-responsive polymer widely used in biosensors for temperature-dependent analyte detection. It enables the controlled release of signaling molecules or reagents upon reaching a specific temperature, enhancing sensor performance and sensitivity.
pH-Sensitive Polymers
pH-responsive polymers are used in drug delivery systems, releasing their payload under specific pH conditions. pH-responsive polymers, such as poly(acrylic acid), are also used in biosensors to detect pH changes, which can indicate the presence of biomolecules (e.g., glucose or lactate) or reflect the state of a biological environment, such as an infection.
Additionally, pH-sensitive polymers are utilized in wound healing materials, including smart dressings that respond to pH variations in the wound environment. As pH levels fluctuate due to infection or healing, these materials can swell, adjust their properties to maintain optimal moisture, or release therapeutic agents as needed.
Bio-Based Monomers
Bio-based monomers are organic compounds derived from renewable biological resources such as plants, animals, and microorganisms, rather than fossil fuels. These monomers contribute to the development of biodegradable and biocompatible polymers, promoting sustainability and reducing the environmental impact associated with traditional petrochemical-derived polymers. Polylactic acid (PLA) is a widely used biodegradable polymer, commonly found in packaging and disposable products. Biocompatible polymers such as polyethylene, and poly(methyl methacrylate) (PMMA) are used in implants, prosthetics, and tissue engineering applications.