Skip to Content
Merck
MMP-13 Substrate, Fluorogenic A quenched fluorescence substrate for MMP-13 (collagenase-3; kcat/Km = 1.09 x 10⁶ M⁻¹s⁻¹).

MMP 13 Substrate, fluorogenic

MMP-13 Substrate, Fluorogenic A quenched fluorescence substrate for MMP-13 (collagenase-3; kcat/Km = 1.09 x 10⁶ M⁻¹s⁻¹).
Synonym(s):
MMP-13 Substrate, Fluorogenic
Empirical Formula (Hill Notation):
C51H65N13O15
Molecular Weight:
1100.14