The Colors of Compatibility
ColorWheel® Flow Cytometry Antibodies and Dyes
The ColorWheel® flow cytometry portfolio utilizes a proprietary technology optimized for flow cytometry that allows users to independently select antibodies and dyes for assembly in any desired combination. With less than 5 minutes of hands-on time, these antibodies and dyes serve as an analog to primary conjugated antibodies. This mix-and-match ability is supplemented with a lyophilized presentation product for enhanced stability and ambient shipping, as well as preservative-free presentation for sustainability and wider sample type compatibility.
The ColorWheel® portfolio of antibodies and dyes provides users with the freedom to design the experiments they want to conduct with the flexibility to adapt today’s experiments to tomorrow’s questions — all in a simple procedure with no compromise in performance. View our simple 3-Step ColorWheel® Flow Cytometry Reagent Preparation Protocol.”
ColorWheel® antibodies and dyes help you obtain reliable, reproducible results by reducing the “unknowns” in your experiment.
Spend less to discover more with this trial offer to get a discount on your first order of ColorWheel® flow cytometry antibodies and dyes. Complete the form by clicking the button below and a representative will be in touch with your discount offer.
Comparison to Conventional Flow Cytometry Methods |
---|
Multiplexing in flow cytometry can be a puzzle that researchers have to piece together. Flow cytometry multiplexing involves balancing various pieces of the puzzle including the configuration of the instrument, balancing antigen expression with dye brightness, and antibody-dye combination availability. If it is not possible to fit all of these factors into one experiment, researchers need to compromise by performing multiple assays. Running multiple assays takes more time and introduces different experimental conditions causing variability that may lead to unreliable data. The flexibility of ColorWheel® technology helps you fit the multiplexing puzzle pieces together.
ColorWheel® antibodies and dyes are lyophilized and provide 5+ years of shelf life. These products can be stored longer than traditional pre-labeled antibodies and dyes, making them ideal for reagent storage at core facilities. While targets can change per researcher, the configuration of the flow cytometers within the facility, and thus the compatible dyes, remain constant. With the ColorWheel® portfolio, this consistency in dye compatibility allows for stocking of the individual dye components that work with the instrument, preventing disruption in reagent supply and taking advantage of bulk discounts. With a steady supply of dye reagent always in stock, simply choose the ColorWheel® antibody of interest, mix and match as needed, and run your assay.
Ensure you always have reagent on hand by purchasing the ColorWheel® dyes that fit your instrument in bulk to save time and money. Purchase bundles of ColorWheel® products and receive an automatic discount in your shopping cart.
For answers to common questions about ColorWheel® products, see our ColorWheel® FAQs page.
Unless otherwise stated in the Product(s) specifications, any Antibody product is sold for internal research use only and may not be used for any other purpose, which includes but is not limited to, any commercial, diagnostic, or therapeutic use. Our validation processes pertain only to research uses and do not confirm or assure that our antibodies can be used for any unauthorized uses as set forth herein.
ColorWheel® technology requires BOTH a ColorWheel® antibody AND a ColorWheel® dye for flow cytometry research applications. The antibody and dye are sold separately.