Expanded MILLIPLEX® Mouse Cytokine Multiplex Panel
Mouse models are essential in biomedical research, particularly in immunology, cancer, and infectious diseases. Measuring cytokines in these models is crucial as they regulate immunity, inflammation, and hematopoiesis. Quantifying cytokine levels with mouse cytokine panels provides insights into immune responses, disease progression, and treatment efficacy, aiding in the development of new therapies and understanding disease mechanisms.
MILLIPLEX® Mouse Cytokine/Chemokine/Growth Factor Expanded Panel 1
Features and Benefits:
- Flexible Formats: The flexibility to configure any combination of up to 68 analytes, custom premixed beads, or choose a fixed 41-plex or 68-plex premix or bulk premixed kit.
- Efficient Sample Management: Analyze multiple targets with just 25 µL/well (1:2 diluted), conserving precious samples and maximizing data output from limited material.
- Incubation Options to Fit Your Schedule: Opt for same-day or overnight incubation to seamlessly integrate into your workflow, enhancing productivity and flexibility.
- Dependable and Reproducible Results: Benefit from proven lot-to-lot consistency, ensuring trustworthy results that are vital for analytical R&D.
- Streamlined Assay Setup: Simplify preparation and reduce the risk of errors with ready-to-use reagents and comprehensive kit components, including Serum Matrix and Quality Controls.
- Enhance Multiplexing Capabilities: Upgrade to the xMAP® INTELLIFLEX® system to fully utilize the 68-plex panel, enabling more comprehensive and efficient data collection, providing deeper insights into complex biological systems.
Mouse Cytokine Panel Details
This MILLIPLEX® kit is available in several formats for your convenience:
| Kit | Analytes |
|---|---|
| Configurable 68-Plex† | Select any of the following analytes – BAFF*, Betacellulin*, CHI3L1*, CXCL16*, EGF, Eotaxin/CCL11, Erythropoietin*, Exodus-2/CCL21, FGF-2, FLT3L*, Fractalkine/CX3CL1*, G-CSF, GDF-15/MIC-1*, GM-CSF, Granzyme B, IFNα, IFNβ, IFNγ, IL-10, IL-11, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-16*, IL-17A, IL-17F, IL-18, IL-1α, IL-1β, IL-2, IL-20*, IL-22, IL-28B/IFNλ3, IL-3, IL-31*, IL-33/NF-HEV (mature), IL-4, IL-5, IL-6, IL-7, IL-9, IP-10/CXCL10, KC/GROα/CXCL1, LIF, LIX/CXCL5, MCP-1/CCL2, MCP-2/CCL8*, MCP-3/CCL7*, MCP-5/CCL12, M-CSF, MDC/CCL22, MIG/CXCL9, MIP-1α/CCL3, MIP-1β/CCL4, MIP-2/CXCL2, MIP-3α/CCL20, MIP-3β/CCL19, RANTES/CCL5, sCD137/4-1BB/TNFRSF9*, sFas/TNFRSF6*, sFasL*, sICAM-1*, sRAGE*, TARC/CCL17, TECK/CCL25*, TNFα, and VEGF-A |
| Fixed 41-Plex Premix | EGF, Eotaxin/CCL11, FGF-2, G-CSF, GM-CSF, IFNα, IFNβ, IFNγ, IL-10, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-17A, IL-17F, IL-18, IL-1α, IL-1β, IL-2, IL-22, IL-28B/IFNλ3, IL-3, IL-33/NF-HEV (mature), IL-4, IL-5, IL-6, IL-7, IL-9, IP-10/CXCL10, KC/GROα/CXCL1, LIF, LIX/CXCL5, MCP-1/CCL2, M-CSF, MIG/CXCL9, MIP-1α/CCL3, MIP-1β/CCL4, MIP-2/CXCL2, RANTES/CCL5, TNFα, and VEGF-A |
| Fixed 68-Plex Premix‡ | BAFF, Betacellulin, CHI3L1, CXCL16, EGF, Eotaxin/CCL11, Erythropoietin, Exodus-2/CCL21, FGF-2, FLT3L, Fractalkine/CX3CL1, G-CSF, GDF-15/MIC-1, GM-CSF, Granzyme B, IFNα, IFNβ, IFNγ, IL-10, IL-11, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-16, IL-17A, IL-17F, IL-18, IL-1α, IL-1β, IL-2, IL-20, IL-22, IL-28B/IFNλ3, IL-3, IL-31, IL-33/NF-HEV (mature), IL-4, IL-5, IL-6, IL-7, IL-9, IP-10/CXCL10, KC/GROα/CXCL1, LIF, LIX/CXCL5, MCP-1/CCL2, MCP-2/CCL8, MCP-3/CCL7, MCP-5/CCL12, M-CSF, MDC/CCL22, MIG/CXCL9, MIP-1α/CCL3, MIP-1β/CCL4, MIP-2/CXCL2, MIP-3α/CCL20, MIP-3β/CCL19, RANTES/CCL5, sCD137/4-1BB/TNFRSF9, sFas/TNFRSF6, sFasL, sICAM-1, sRAGE, TARC/CCL17, TECK/CCL25, TNFα, and VEGF-A |
Highlighted MILLIPLEX® Mouse Multiplex Panels
Response not successful: Received status code 500
Luminex® Instruments and Consumables
Response not successful: Received status code 500
For Research Use Only. Not For Use In Diagnostic Procedures.
Request Info on MILLIPLEX® Mouse Cytokine Expanded Panel 1
Fields with an * are required.
Related Resources
- Article: How to Select Your Immunoassay Platform
Explore how to select the best immunoassay platform for your research by choosing between multiplex and singleplex biomarker detection assays, immunoassay instrument platforms, immunoassay software, and services.
- Article: Overview of Luminex® Multiplex Assay Technology
Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.
- Article: Quality is Built into Every MILLIPLEX® Multiplex Kit
Learn more about how you can rely on the quality we build into each MILLIPLEX® multiplex kit to produce results you trust.
MILLIPLEX® Configurator Tool Update Demo Video
Highlights how easy it is to configure MILLIPLEX® multiplex panels with the updated configurator tool. Order in just 3 steps.