Skip to Content
MilliporeSigma

Skip To

940925

Lithium nickel manganese cobalt oxide

NMC811 cathode active material, battery grade

Synonym(s):

NCM811

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 g
Please contact Customer Service for Availability
$265.00
$198.75
1 kg
Please contact Customer Service for Availability
$942.00

About This Item

Empirical Formula (Hill Notation):
LiNi0.8Mn0.1Co0.1O2
CAS Number:
Molecular Weight:
97.28
MDL number:
NACRES:
NA.21
UNSPSC Code:
12352303

$198.75

List Price$265.00Save 25%
Web-Only Promotion

Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

battery grade

Quality Segment

description

first cycle efficiency: 86-90% (typical)

form

powder

particle size

10 μm (D50, typical)

capacity

(First cycle discharge capacity: >195 mAh/g at 0.1C (typical) >185 mAh/g at 1C (typical))

density

2.15 g/cm3 (lit.)

application(s)

battery manufacturing

SMILES string

[Li].[O].[Mn].[Co].[Ni]

InChI

1S/Co.Li.Mn.Ni.O

InChI key

VGYDTVNNDKLMHX-UHFFFAOYSA-N

General description

Our NMC811 is a high-performance cathode material widely used in rechargeable lithium-ion batteries. It is composed of lithium, nickel, manganese, cobalt, and oxygen, with a nominal composition of 80% nickel, 10% manganese, and 10% cobalt. Our NMC811 has a minimum nickel content of 80.0% of the metal content (≥48.2 wt% overall), translating to a nickel-rich composition between NMC811 and NMC85:05:10. This unique composition gives NMC811 its exceptional energy density and power capabilities. The material adopts a layered crystal structure, which facilitates the intercalation of lithium ions between the layers during charge and discharge cycles, enabling efficient energy storage and release.

Application

Our battery grade NMC811 is specifically engineered for advanced lithium-ion battery applications, such as electric vehicles, energy storage systems, and portable electronic devices. It offers an exceptional balance of high energy density and power capability, making it an ideal choice for manufacturers seeking to enhance the performance of their lithium-ion battery products. With an initial discharge capacity exceeding 188 mAh/cm³ at 1C rate and a first discharge efficiency greater than 88%, our NMC811 delivers outstanding energy storage and power output. Additionally, the synthesis process of our NMC811 ensures a controlled particle size distribution (D50 of 9-12 microns) and a low surface area (< 1 m²/g), enabling fast kinetics and minimizing surface degradation. The high tap density, typically around 2 g/cm³, allows for the formation of denser electrode films, further increasing the overall energy density of the battery.


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library