Iupac Name
7-bromo-2H-chromen-2-one
Inchi Code
1S/C9H5BrO2/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5H
InChI key
PFJPBLFPJISXMF-UHFFFAOYSA-N
storage temp.
Sealed in dry, room temperature
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/19063-56-0.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/19063-56-0.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service