Skip to Content
MilliporeSigma

COMH95E96E31

Combi-Blocks, Inc.

Glutathione oxidized, hydrate

Synonym(s):

(2S)-2-amino-5-({(1R)-1-[({(2R)-2-{[(4S)-4-amino-4-carboxybutanoyl]amino}-3-[(carboxymethyl)amino]-3-oxopropyl}disulfanyl)methyl]-2-[(carboxymethyl)amino]-2-oxoethyl}amino)-5-oxopentanoic acid hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
630.65
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H32N6O12S2.H2O/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38;/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38);1H2/t9-,10-,11-,12-;/m0./s1

Inchi Key

AQWLIFJHVLBNMB-AAQZBYNGSA-N

Storage temp.

Freezer

Purity

95%

Country of Origin

CN



MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QH-2597



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library