1.08815

SAFC

Zinc chloride

EMPROVE® API Ph Eur,BP,USP

Synonym(s):
Zinc dichloride, Zinc chloride
Empirical Formula (Hill Notation):
Cl2Zn
CAS Number:
Molecular Weight:
136.30
MDL number:
EC Index Number:
231-592-0

storage conditions

Store at +2°C to +25°C.

Quality Level

Agency/Method

BP
Ph. Eur.
USP

vapor pressure

1.33 hPa ( 428 °C)

product line

EMPROVE® API

potency

350 mg/kg LD50, oral (Rat)

pH

5 (20 °C, 100 g/L in H2O)

bp

732 °C/1013 hPa

mp

287-304 °C

density

2.93 g/cm3 at 22 °C

bulk density

1400‑1800 kg/m3

storage temp.

2-25°C

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs. As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Zinc chloride is used in mouth hygiene, as trace element in clinical nutrition, insulin and infusion solutions. Zinc Chloride Emprove® API is manufactured according to ICH Q7 guidelines. A CEP is available.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

hazcat

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

storage_class_code

8B - Non-combustible, corrosive hazardous materials

WGK Germany

WGK 3

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificate of Analysis

Certificate of Origin

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials.

Elemental Impurities Information

Available by subscription or for a fee

Elemental Impurities Information

Available by subscription or for a fee

API Information Package

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service