Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

116765

Sigma-Aldrich

Adenosine A 2A/A1 Receptor Antagonist

The Adenosine A2A/A₁ Receptor Antagonist controls the biological activity of Adenosine A2A/A₁ Receptor. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

Adenosine A 2A/A1 Receptor Antagonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22N4O3 · 2HCl
CAS Number:
Molecular Weight:
475.37
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light yellow

solubility

DMSO: 10 mg/mL, clear, yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

NC(N=C1C2=CC=CC=C2)=NC(C3=C4C=CC(OCCN5CCOCC5)=C3)=C1C4=O

General description

A blood-brain barrier-permeant indenopyrimidinone that acts as a dual A1/A2A antagonist (Ki = 48.2 and 6.5 nM, respectively, against agonist-induced cAMP response in A1- or A2A-expressing CHO-K1 cells) and effectively reverses D2 antagonist haloperidol- (1 mg/kg; s.c.) induced catalepsy in both rats and mice (ED50 = 0.3 and <0.1 mg/kg, respectively) in vivo by simultaneously reversing A1-mediated inhibition of DA (Dopamine) release and A2A-dependent inhibition of D2 receptor response to DA.
The Adenosine A2A/A₁ Receptor Antagonist controls the biological activity of Adenosine A2A/A₁ Receptor. This small molecule/inhibitor is primarily used for Biochemicals applications.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Shook, B.C., et al. 2012. J. Med. Chem.55, 1402.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service