116814
Adenosine 3′,5′-cyclic Monophosphorothioate, Rp-Isomer, Triethylammonium Salt
A cell-permeable and reversible inhibitor of protein kinase A (Ki = 11 µM).
Synonym(s):
Adenosine 3′,5′-cyclic Monophosphorothioate, Rp-Isomer, Triethylammonium Salt, Adenosine 3ʹ,5ʹ-cyclic Phosphorothioate-Rp, Rp-cAMPS, TEA, PKA Inhibitor V
Select a Size
$368.00
Select a Size
About This Item
$368.00
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
crystalline solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
solubility
water: 20 mg/mL
shipped in
wet ice
storage temp.
−20°C
InChI
1S/C10H12N5O5PS.C6H15N/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(16)7-4(19-10)1-18-21(17,22)20-7;1-4-7(5-2)6-3/h2-4,6-7,10,16H,1H2,(H,17,22)(H2,11,12,13);4-6H2,1-3H3/t4-,6-,7-,10-,21-;/m1./s1
InChI key
OXIPZMKSNMRTIV-RNSTXJOVSA-N
General description
Biochem/physiol Actions
PKA
Warning
Reconstitution
Other Notes
Van der Kloot, W., and Branisteanu, D.D. 1992. Eur. J. Phys.420, 336.
Yokozaki, H., et al. 1992. Cancer Res.52, 2504.
Butt, E., et al. 1990. FEBS Lett.263, 47.
Dostmann, W.R., et al. 1990. J. Biol. Chem.265, 10484.
Rothermel, J.D. 1988. Biochem. J. 251, 757.
Eckstein, F. 1985. Annu. Rev. Biochem.54, 367.
de Wit, R.J., et al. 1984. Eur. J. Biochem.142, 255.
O′Brian, C.A., et al. 1982. Biochemistry21, 4371.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service