MilliporeSigma
All Photos(1)

Documents

203119

Sigma-Aldrich

Innolink Biotin 354S

Synonym(s):
NHS-Chromogenic Biotin
Empirical Formula (Hill Notation):
C38H50N8O10S
Molecular Weight:
810.92

Assay

≥90% (reversed phase HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

DMF: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
809608B205912-539
Innolink Biotin 354S

Sigma-Aldrich

203119

Innolink Biotin 354S

form

solid

form

powder

form

powder

form

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Upstate®

storage condition

OK to freeze

storage condition

-

storage condition

-

storage condition

-

solubility

DMF: 50 mg/mL

solubility

-

solubility

H2O: 10 mg/mL

solubility

-

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

wet ice

General description

A modified biotin conjugate in which biotin is linked to a bis-aryl hydrazone chromophore and aromatic succinimidyl ester via a long-chain PEG4 linker. The bis-aryl hydrazone chromophore allows for direct spectrophotometric quantitation of total incorporated biotin. The long-chain PEG4 linker preserves biotin/avidin affinity as well as increases solubility. The aromatic succinimidyl ester allows for higher efficiency modification of amines in aqueous buffers.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, purge with inert gas, aliquot, and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Steinberg-Tatman, G., et al. 2006. Bioconjug. Chem.17, 841.
Kline, M.C., et al. 2005. J. Forensic Sci.50, 1.
Kozlov, I.A., et al. 2004. Biopolymers73, 621.
Chen, Y., et al. 2004. Chem. Commun.24, 2804.
Kozlov, I.A., et al. 2004. Biopolymers73, 621.
Zhong, X.B., et al. 2003. Proc. Natl. Acad. Sci. USA100, 11559.
Hartmann, D.M., et al. 2002. Journal of Materials Research17, 473.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service