Skip to Content
MilliporeSigma

217696

Sigma-Aldrich

Cdk1 Inhibitor

CGP74514A

Synonym(s):

Cdk1 Inhibitor

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$394.00

$394.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$394.00

About This Item

Linear Formula:
C19H24ClN7
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

$394.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

(Off-white solid)

solubility

soluble (DMSO (10 mg/ml))

storage temp.

−20°C

General description

A cell-permeable, potent, reversible, ATP-competitive and selective inhibitor of Cdk1/cyclin B (IC50 = 25 nM). Affects the activities of other kinases only at higher concentrations (IC50 = 6.1 µM for PKCα, 125 µM for PKA, and >10 µM for EGFR). Also reported to induce mitochondrial damage and apoptosis in several human leukemia cell lines (>3 µM). At lower concentrations (~1 µM) an initial G2M cell cycle arrest and eventual apoptosis has been reported to occur in U937 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service