Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

218850

Sigma-Aldrich

Casein Kinase I Inhibitor V, LH846

The Casein Kinase I Inhibitor V, LH846 controls the biological activity of Casein Kinase I. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Casein Kinase I Inhibitor V, LH846, N-(5-Chloro-6-methylbenzo[d]thiazol-2-yl)-2-phenylacetamide, N-(5-Chloro-6-methyl-1,3-benzothiazol-2-yl)-2-phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClN2OS
CAS Number:
Molecular Weight:
316.81
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC1=CC2=C(SC(NC(CC3=CC=CC=C3)=O)=N2)C=C1C

InChI

1S/C16H13ClN2OS/c1-10-7-14-13(9-12(10)17)18-16(21-14)19-15(20)8-11-5-3-2-4-6-11/h2-7,9H,8H2,1H3,(H,18,19,20)

InChI key

DYHAMRNAHTWYKY-UHFFFAOYSA-N

General description

A cell-permeable benzothiazolo compound that acts a potent, selective, and reversible inhibitor of CKIδ (IC50 = 290 nM). Exhibits much reduced potency towards CKIε and CKIα isoforms (IC50 =1.3 and 2.5 µM, respectively). Does not affect the activity of CK2 or other kinases in a 59-protein kinase panel. Shown to block CKIδ-dependent period protein PER1 phosphorylation and diminish its proteasomal degradation in a dose-dependent manner. Also, shown to lengthen the circadian period in U2OS cells (10 h at 8 µM) without significantly affecting the amplitude of Per2-dLuc and Bmal1-dLuc rhythms.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lee, J.W., et al. 2011. Angew. Chem. Int. Ed.50, 10608.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service