Skip to Content
MilliporeSigma

219498

Sigma-Aldrich

Cathepsin S Substrate, Fluorogenic

An internally quenched, highly cathepsin S-selective, fluorogenic peptide substrate that is efficiently cleaved by CatS, but not by Cathepsin B & L.

Synonym(s):

Cathepsin S Substrate, Fluorogenic, Mca-GRWPPMG~LPWEK(Dnp)- D-R-NH2, Mca-GRWPPMG~LPWEK(Dnp)-D-R-NH2

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C93H123N25O23S
Molecular Weight:
1991.19
UNSPSC Code:
12352200

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

An internally quenched, highly cathepsin S-selective, fluorogenic peptide substrate that is efficiently cleaved by CatS (kcat/Km = 1.018 µM-1 S-1), but not by Cathepsin B & L (kcat/Km = 0.004 and 0.032 µM-1 S-1, respectively). Studies using CatS-specific inhibitor LHVS reveal that the substrate is not cleaved by other proteolytic enzymes, including CatB, CatH, CatL, and CatX, present in endosomal extracts from human B cell line WT100, dendritic cells, and macrophages (excitation maximum: 340 nm; emission maximum: 405 nm).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Mca-Gly-Arg-Trp-Pro-Pro-Met-Gly~Leu-Pro-Trp-Glu-Lys(Dnp)-D-Arg-NH₂

Other Notes

Lutzner, N., and Kalbacher, H. 2008. J. Biol. Chem.283, 36185.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service