Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239831

Millipore

D-Cycloserine

D Cycloserine is a partial agonist of NMDA receptors at the glycine modulatory site. Reported to improve memory-dependent tasks in rats. Neutral and acidic solutions are unstable.

Synonym(s):

D-Cycloserine, D-4-Amino-3-isoxazolidinone, Oxamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2O2
CAS Number:
Molecular Weight:
102.09
MDL number:
UNSPSC Code:
12352209

Quality Level

Assay

≥95% (HPLC)

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white to pale yellow

solubility

water: 50 mg/mL
aqueous buffer, pH 10: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C3H6N2O2/c4-2-1-7-5-3(2)6/h2H,1,4H2,(H,5,6)

InChI key

DYDCUQKUCUHJBH-UHFFFAOYSA-N

General description

Partial agonist of NMDA receptors at the glycine modulatory site. Reported to improve memory-dependent tasks in rats. Neutral and acidic solutions are unstable.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, refrigerate (4°C). Alkaline stock solutions (pH 10.0) are stable up to 1 week at 4°C.

Other Notes

Pussinen, R., and Sirvio, J. 1999. J. Psychopharmacol.13, 171.
Tsai, G.E., et al. 1999. Am. J. Psychiatr. 156, 467.
Baran, H., et al. 1994. Brain Res. 652, 195.
Hood, W.F., et al. 1989. Neurosci. Lett.98, 91.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service