Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

260930

Sigma-Aldrich

DNA Polymerase Sliding Clamp Inhibitor, RU7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
solubility:
water: soluble
Pricing and availability is not currently available.

solubility

water: soluble

Quality Level

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service