346104
Plk2, GST- Fusion, Human, Recombinant, S. frugiperda
Synonym(s):
Polo-like kinase 2, Human, Recombinant
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
Recommended Products
Assay
≥70% (SDS-PAGE)
Quality Level
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Full length, recombinant, human Plk2 fused at the N-terminus to GST and expressed in S. frugiperda insect cells using a baculovirus expression system. Plk2 is a G1 specific kinase that is specifically detected in brain, heart, and lung. Plk2 expression increases in the presence of DNA-damaging agents. The active kinase is useful for the study of Plk2 regulation and inhibitor screening.
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Unit Definition
One unit is defined as the amount of enzyme that will phosphorylate 1 pmol SER/THR 16 substrate per min at 30°C, pH 7.5.
Physical form
In 100 mM NaCl, 25 mM Tris-HCl, 3 mM DTT, 50% glycerol, 0.05% Tween®-20 detergent, pH 8.0.
Reconstitution
Following initial thaw, aliquot and freeze (-70°C).
Other Notes
Eckerdt, F., et al. 2005. Oncogene24, 267.
Warnke, S., et al. 2004.Curr Biol.14, 1200.
Warnke, S., et al. 2004.Curr Biol.14, 1200.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service