Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

361567

Sigma-Aldrich

GSK-3β Inhibitor XXIV

The GSK-3β Inhibitor XXIV, also referenced under CAS 1005201-24-0, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3β Inhibitor XXIV, 2-Methyl-5-(3-[4-(methylsulfinyl)phenyl)-1-benzofuran-5-yl)-1,3,4-oxadiazole, Racemic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2O3S
CAS Number:
Molecular Weight:
338.38
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

CS(C(C=C1)=CC=C1C2=COC3=C2C=C(C4=NN=C(C)O4)C=C3)=O

InChI

1S/C18H14N2O3S/c1-11-19-20-18(23-11)13-5-8-17-15(9-13)16(10-22-17)12-3-6-14(7-4-12)24(2)21/h3-10H,1-2H3

InChI key

RCKYSTKYIVULEK-UHFFFAOYSA-N

General description

The racemic mixture of a cell-permeable oxadiazolo-benzofuranylphenylsulfoxide that inhibits GSK-3β (IC50 = 35, 34, and 140 nM using the racemate, the (S), and the (R) enantiomer, respectively) in a highly selective manner as evidenced by its eutomer′s inability to inhibit 10 other serine/threonine and 12 other tyrosine kinases even at concentrations as high as 10 µM. Reported to exhibit good aqueous solubility (38 µg racemate/ml pH 6.8 buffer) and pharmacokinetic properties, being orally active in both rats and mice in vivo (Plasma AUC0-24h = 1380.6 and 457 ng•h/mL, respectively; 3 mg eutomer/kg via p.o.). The eutomer is also shown to be blood-brain-permeant in mice (brain AUC0-24h = 734 ng•h/mL; 3 mg/kg via p.o.) and oral dosing 30 min prior to CWS (cold water stress; 4 min below-neck inversion in 1-2 °C water) is demonstrated to reduce brain tau Thr205 phosphorylation (by 35%; 10 min post CWS) in mice in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Saitoh, M., et al. 2009. J. Med. Chem.52, 6270.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service