Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

361570

Sigma-Aldrich

GSK-3β Inhibitor XXVII

The GSK-3β Inhibitor XXVII controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3β Inhibitor XXVII, 3-Amino-6-(4-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-N-(pyridin-3-yl)pyrazine-2-carboxamide, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23N7O3S · 2.5HCl · 5H2O
Molecular Weight:
634.75
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable pyridinylpyrazinecarboxamide compound that acts as a highly potent, reversible, and ATP-competitive inhibitor of GSK-3β (Ki = 4.9 nM) with ~110-fold greater selectivity over Cdk2 (Ki = 540 nM). Shows only a trivial effect on the activities of 26-related kinases at much higher doses. Blocks GSK-3β-mediated phosphorylation of Ser396 in stably transfected 3T3 fibroblasts expressing four-repeat tau protein (IC50 = 76 nM). Readily crosses blood-brain barrier and exhibits desirable bioavailability and aqueous solubility.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GSK-3β
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Berg. S., et al. 2012. J. Med. Chem.55, 9107.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service