Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

368620

Sigma-Aldrich

Group III Caspase Inhibitor I

The Group III Caspase Inhibitor I controls the biological activity of Group III caspases (caspase-6, -8, -9, and -10). This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Group III Caspase Inhibitor I, Z-AE(OMe)VD(OMe)-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H39FN4O10
Molecular Weight:
610.63
UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: 20 mM

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable, and irreversible inhibitor of Group III caspases (caspases-6, -8, -9, and -10), although more effective towards caspases -6 and -8. Also inhibits caspase-1 and caspase-3. When using with purified native or recombinant enzyme, pretreatment with an esterase is required.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-1, caspase-3, caspase-6, caspase-8, caspase-9, caspase-10
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Ala-Glu(OMe)-Val-Asp(OMe)-CH₂F*

Reconstitution

Following reconstituion, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Analysis Note

Single spot by TLC

Other Notes

Garcia-Calvo, M., et al. 1998. J. Biol. Chem. 273, 32608.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service