371731
Anti-Gsα-Subunit Rabbit pAb
liquid, Calbiochem®
Select a Size
Select a Size
About This Item
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
form
liquid
contains
≤0.1% sodium azide as preservative
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
dilution
(ELISA (1:1000)
Immunoblotting (1:1000))
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
1 of 4
This Item | 371754 | 345757 | 178479 |
---|---|---|---|
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
biological source rabbit | biological source rabbit | biological source rabbit | biological source goat |
antibody form serum | antibody form purified antibody | antibody form serum | antibody form serum |
clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
storage temp. −20°C | storage temp. −70°C | storage temp. −20°C | storage temp. −20°C |
General description
Immunogen
Application
Warning
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service