Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

373275

Sigma-Aldrich

Hh Signaling Antagonist XII, HPI-1

The Hh Signaling Antagonist XII, HPI-1, also referenced under CAS 599150-20-6, controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Hh Signaling Antagonist XII, HPI-1, 2-Methoxyethyl-4-(3-hydroxyphenyl)-7-(2-methoxyphenyl)-2-methyl-5-oxo-4,6,7,8-tetrahydro-1H-quinoline-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO6
CAS Number:
Molecular Weight:
463.52
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C27H29NO6/c1-16-24(27(31)34-12-11-32-2)25(17-7-6-8-19(29)13-17)26-21(28-16)14-18(15-22(26)30)20-9-4-5-10-23(20)33-3/h4-10,13,18,25,28-29H,11-12,14-15H2,1-3H3

InChI key

ZXSWZQSYZYMZKS-UHFFFAOYSA-N

General description

A cell-permeable oxo-tetrahydroquinolinecarboxylate compound that acts as a downstream Hh (Hedgehog) pathway blocker, effectively inhibiting Gli-dependent transcription activity in both Sufu+/+ NIH 3T3 (shh-LIGHT2) and Sufu-/- fibroblast cultures (IC50 = 1.5 & 3 µM, respectively) in a manner similar to two other Hh downstream blockers, HPI-3 (Cat. No. 373276) and GANT-61 (Cat. No. 373401). However, only HPI-1, but not HPI-3 or GANT-61 (no effect at 30 µM), remains effective against Sufu-/- fibroblasts overexpressing Gli1 or Gli2 (IC50 = 6 & 4 µM, respectively). HPI-1 is also shown to inhibit the ligand-independent Hh pathway activation in SmoM2-expressing primary murine CGNPs (Cerebellar Granule Neuron Precursors) and NIH 3T3 (smoM2-LIGHT) cultures.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hyman, J.M., et al. 2009. Proc. Natl. Acad. Sci. USA106, 14132.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service