373385
Hh Signaling Antagonist VII, JK184
The Hh Signaling Antagonist VII, JK184, also referenced under CAS 315703-52-7, controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.
Synonym(s):
Hh Signaling Antagonist VII, JK184, N-(4-Ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)thiazol-2-amine
About This Item
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
pink
solubility
methanol: 10 mg/mL
DMSO: 40 mg/mL
shipped in
wet ice
storage temp.
−20°C
InChI
1S/C19H18N4OS/c1-3-24-15-9-7-14(8-10-15)21-19-22-16(12-25-19)18-13(2)20-17-6-4-5-11-23(17)18/h4-12H,3H2,1-2H3,(H,21,22)
InChI key
ROYXIPOUVGDTAO-UHFFFAOYSA-N
General description
Packaging
Warning
Reconstitution
Other Notes
Lee, J., et al. 2007. ChemBioChem.8, 1916.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service