Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420118

Sigma-Aldrich

JNK Inhibitor I, (L)-Form, Cell-Permeable, Negative Control

The JNK Inhibitor I, (L)-Form, Cell-Permeable, Negative Control controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK Inhibitor I, (L)-Form, Cell-Permeable, Negative Control, (L)-HIV-TAT₄₈₋₅₇-PP, GRKKRRQRRRPP-NH₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H124N34O13
Molecular Weight:
1589.90
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 2 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A highly cell-permeable 10-amino acid carrier peptide derived from HIV-TAT48-57 sequence that is modified with two proline residues. Shown to accumulate into a variety of cells. Serves as a useful negative control for studies involving JNK Inhibitor I, (L)-Form, Cell-Permeable (Cat. No. 420116).
A highly cell-permeable carrier decapeptide derived from HIV-TAT48-57 sequence that is modified with two proline residues. Serves as a useful negative control for studies involving JNK Inhibitor I, (L)-Form, Cell-permeable (Cat. No. 420116).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
A useful negative control for studies involving JNK Inhibitor I, (L)-Form, Cell-Permeable
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Pro-Pro-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bonny, C., et al. 2001. Diabetes50, 77.
Vocero-Akbani, A. M., et al 1999. Nat. Med.5, 29.
Schwarze, S. R., et al. 1999. Science285, 1573.
Nagahara, H., et al. 1998. Nat. Med.4, 1449.
Vives, E., et al. 1997. J. Biol. Chem.272, 16010.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service