Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420140

Sigma-Aldrich

JNK Inhibitor X, BI-78D3

The JNK Inhibitor X, BI-78D3, also referenced under CAS 883065-90-5, controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK Inhibitor X, BI-78D3, 4-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-5-(5-nitrothiazol-2-ylthio)-4H-1,2,4-triazol-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9N5O5S2
CAS Number:
Molecular Weight:
379.37
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C13H9N5O5S2/c19-11-15-16-12(25-13-14-6-10(24-13)18(20)21)17(11)7-1-2-8-9(5-7)23-4-3-22-8/h1-2,5-6H,3-4H2,(H,15,19)

InChI key

QFRLDZGQEZCCJZ-UHFFFAOYSA-N

General description

A cell-permeable benzodioxin-triazole compound that competes with JNK-interacting protein-1 (IC50 = 500 nM in competitive binding to JNK1 with pepJIP1) and D-domain-containing substrates binding (Ki = 200 nM in competitive binding to JNK1 with ATF2) by targeting JNK JIP1-interacting site (Kd = 8.1 µM). BI-78D3 inhibits the phosphorylation of ATF2, but not that of a D-domain-less peptide substrate, by JNK1 in kinase assays, while showing 100-fold less potency against p38α and no effect against mTOR or PI3-Kα. Shown to inhibit the TNF-α-stimulated GFP-cJun phosphorylation in transfected HeLa cultures in vitro (IC50 = 12.4 µM) and effectively reduce ConA-induced murine liver failure (10 mg/kg, i.p.) as well as restore insulin sensitivity in a murine type 2 diabetes model (25 mg/kg, i.p.) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Stebbins, J.L., et al. 2008. Proc. Natl. Acad. Sci. USA105, 16809.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service