Sign In to View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C23H20ClN3O
CAS Number:
Molecular Weight:
389.88
UNSPSC Code:
12352200
Skip To
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 100 mg/mL
shipped in
ambient
storage temp.
2-8°C
General description
A cell-permeable indole-3-propanamide immunosuppressant that is shown to selectively inhibit the kinase activity of JAK3 (by 81% and 36% at 90 and 30 µM, respectively) over that of JAK2 (by 29% and 0% at 90 and 30 µM, respectively) and reduce JAK1/3-dependent phosphorylations of Akt, STAT5a/b, and Erk1/2 in IL-2-stimulated CTL-L2 cells, but not JAK1/2-dependent STAT1 phosphorylation in INF-γ-stimulated U266 cultures. Reported to inhibit ConA-stimulated murine splenocytes and PHA-stimulated human PBL proliferation (IC50 = 17 and 25 µM, respectively) in vitro and be efficacious in ameliorating delayed hypersensitivity reaction in mice (by ~78% with a daily oral dose of 50 mg/kg) and in prolonging the survival of heart transplant-recipient rats (by >3-fold with 60 mg/kg/day, p.o.) in vivo.
Packaging
Packaged under inert gas
Other Notes
Carbonnelle, D., et al. 2009. J. Pharm. Exp. Ther.331, 710.
Carbonnelle, D., et al. 2007. Eur. J. Med. Chem.42, 686.
Carbonnelle, D., et al. 2007. Eur. J. Med. Chem.42, 686.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service