420147
JNK Inhibitor XIV, SR-3306
The JNK Inhibitor XIII, SR-3306 controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
JNK Inhibitor XIV, SR-3306, N-(4-(3-(6-Methylpyridin-3-yl)-1H-1,2,4-triazol-1-yl)phenyl)-4-(3-morpholinophenyl)pyrimidin-2-amine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C28H26N8O
Molecular Weight:
490.56
UNSPSC Code:
12352200
Quality Level
Assay
≥95% (HPLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
pale yellow
solubility
DMSO: 10 mg/mL
shipped in
ambient
storage temp.
2-8°C
General description
A cell-permeable, blood-brain-barrier-permeant, and orally available anilinopyrimidine compound that acts as a highly selective, potent, reversible, and ATP-competitive JNK inhibitor (IC50 = 67, 283, and 159 nM against JNK1, JNK2, and JNK3, respectively), exhibiting little or no activity toward a panel of more than 300 kinases, including p38 (IC50 >20 µM). Shown to inhibit c-jun phosphorylation both in STZ- (Streptozotocin, Cat. No. 572201) stimulated INS-1 β-pancreatic cells in vitro (IC50 = 216 nM) and in SNpc (substantia nigra pars compacta) TH- (tyrosine hydroxylase) positive neurons (by 75%; single p.o. dose of 30 mg/kg) from MPTP- (N-methyl-4-phenyl-1,2,3,6-tetrahydropyridine) treated mice in vivo. Effectively protects against SNpc TH-positive dopaminergic neuron loss in mice (46% vs. 27% loss with vehicle or 30 mg/kg SR-3306; p.o. b.i.d.) and rats (6-fold increase of vehicle control cell count; 10 mg/kg/day; s.c.) due to MPTP and 6-OHDA- (6-hydroxydopamine) intoxication, respectively, in vivo.
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Other Notes
Chambers, J.W., et al. 2011. ACS Chem Neurosci.2, 198.
Crocker, C.E., et al. 2011. ACS Chem. Neurosci.2, 207.
Crocker, C.E., et al. 2011. ACS Chem. Neurosci.2, 207.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service