Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

420175

Sigma-Aldrich

Cancer Hydrolase KIAA1363 Inhibitor, JW480

Synonym(s):

Cancer Hydrolase KIAA1363 Inhibitor, JW480, 2ʹ-Isopropylphenyl-2-(2-naphthyl)ethylcarbamate, AADACL1 Inhibitor, Arylacetamide Deacetylase-Like 1 Inhibitor, NCEH1 Inhibitor, Neutral Cholesterol Ester Hydrolase 1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23NO2
CAS Number:
Molecular Weight:
333.42
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(OC1=CC=CC=C1C(C)C)NCCC2=CC3=C(C=C2)C=CC=C3

InChI

1S/C22H23NO2/c1-16(2)20-9-5-6-10-21(20)25-22(24)23-14-13-17-11-12-18-7-3-4-8-19(18)15-17/h3-12,15-16H,13-14H2,1-2H3,(H,23,24)

InChI key

PCNJGBMWAZRVEA-UHFFFAOYSA-N

General description

A membrane-permeable carbamate that acts as a highly potent and selective inhibitor against the 2-acetyl MAGE (monoakylglycerol ether) hydrolase KIAA1363/AADACL1 of both human (IC50 = 12 nM; PC3 cell lysate) and murine (IC50 = 20 nM; mouse brain membrane homogenate) origin, presumably via covalent/irreversible carbamoylation of enzyme active site nucleophilic serine, with no significant effect toward other PC3 or murine brain serine hydrolase activities, including HSL, FAAH, and AChE. Readily crosses blood-brain barrier in mice (complete inhibition of brain KIAA1363 in 4 h after single i.p. or o.g./oral gavage dose of 5 mg/kg and 20 mg/kg, respectively). Reported to inhibit the survival of PC3 prostate cancer cells in cultures in vitro (by 34%; 48 h; 1 µM) and suppress PC3-derived tumor growth in mice in vivo (by 56% on day 33; 80 mg/kg/day; o.g.). Also inhibits blood carboxylesterase.
A membrane-permeable, highly potent and selective inhibitor against the 2-acetyl MAGE (monoakylglycerol ether) hydrolase KIAA1363/AADACL1 of both human (IC50 = 12 nM; PC3 lysate) and murine (IC50 = 20 nM; brain membrane samples) origin, with no significant effect toward other PC3 or murine brain serine hydrolase activities, including HSL, FAAH, and AChE. Readily crosses blood-brain barrier in mice and effectively suppresses PC3-derived tumor growth in mice in vivo (by 56% on day 33; 80 mg/kg/day; o.g.). Also inhibits blood carboxylesterase.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Chang, J.W., et al. 2011. Chem. Biol.18, 476.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service