Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444236

Sigma-Aldrich

Anti-MMP-9 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Gelatinase B, Anti-Matrix Metalloproteinase 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, marmoset

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... MMP9(4318)

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes all forms of MMP-9 under reducing and non-reducing conditions.
Recognizes all forms of MMP-9 under reducing and non-reducing conditions. Exhibits only slight cross-reactivity with MMP-2. Does not cross-react with MMP-1, MMP-3, or MMP-8.
This Anti-MMP-9 Rabbit pAb is validated for use in ELISA, Immunoblotting, Immunofluorescence, Immunohistochemistry, Immunoprecipitation for the detection of MMP-9.

Immunogen

human pro-MMP-9

Application

ELISA (see comments)

Immunoblotting (1:500-1:1000)

Immunofluorescence (1:50-1:250)

Immunohistochemistry (1:40-1:150)

Immunoprecipitation (1:20-1:60)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Exhibits very slight reactivity with MMP-2. Does not cross-react with collagenases, MMP-1, MMP-3, or MMP-8. This antibody has also been reported to work for ELISA. Variables associated with assay conditions will dictate the proper working dilution.
Kolkenbrock, H., et al. 1996. Biol. Chem.377, 529.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Diessler et al.
Reproduction in domestic animals = Zuchthygiene, 52(1), 35-43 (2016-11-20)
The zonary and endotheliochorial dog placenta is the most invasive placenta of carnivores. The importance of matrix metalloproteinases (MMP) in placenta invasiveness has been determined in several mammals including species with haemochorial, epitheliochorial and endotheliochorial placentation. Regarding the latter, the
Vignesh Venkatakrishnan et al.
The Journal of biological chemistry, 295(36), 12648-12660 (2020-07-16)
Protein glycosylation is essential to trafficking and immune functions of human neutrophils. During granulopoiesis in the bone marrow, distinct neutrophil granules are successively formed. Distinct receptors and effector proteins, many of which are glycosylated, are targeted to each type of
Nicolas T Kummer et al.
Journal of cellular biochemistry, 113(6), 1998-2008 (2012-01-19)
Arachidonate 5-lipoxygenase (ALOX5) expression and activity has been implicated in tumor pathogenesis, yet its role in papillary thyroid carcinoma (PTC) has not been characterized. ALOX5 protein and mRNA were upregulated in PTC compared to matched, normal thyroid tissue, and ALOX5
Anna Michopoulou et al.
Matrix biology : journal of the International Society for Matrix Biology, 94, 1-17 (2020-07-06)
Re-epithelialization describes the resurfacing of a skin wound with new epithelium. In response to various stimuli including that of growth factors, cytokines and extracellular matrix (ECM), wound edge epidermal keratinocytes undergo cytoskeleton rearrangements compatible with their motile behavior and develop
Robyn Nagel et al.
Parasites & vectors, 8, 453-453 (2015-09-17)
Blastocystis species are common enteric human parasites and carriage has been linked to Irritable Bowel Syndrome (IBS), particularly diarrhoea-predominant IBS. The spectrum of immune reactivity to Blastocystis proteins has been reported previously in symptomatic patients. We investigated differences in serum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service