Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444965

Sigma-Aldrich

MEK Inhibitor VIII

The MEK1/2 Inhibitor II, also referenced under CAS 212631-61-3, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

MEK Inhibitor VIII, N-Cyclopropylmethoxy-3,4,5-trifluoro-2-(4-iodo-2-methylphenylamino)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16F3IN2O2
CAS Number:
Molecular Weight:
476.23
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 45 mg/mL
ethanol: 45 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H16F3IN2O2/c1-9-6-11(22)4-5-14(9)23-17-12(7-13(19)15(20)16(17)21)18(25)24-26-8-10-2-3-10/h4-7,10,23H,2-3,8H2,1H3,(H,24,25)

InChI key

UHAXDAKQGVISBZ-UHFFFAOYSA-N

General description

A cell-permeable and orally available amino-benzamide compound that acts as a potent and non-ATP-competitive inhibitor of MEK1/2 (IC50 = 8 nM) with an excellent selectivity over ERK, c-Src, Cdk′s, and PI 3-Kγ (IC50 >1.0 µM). Reported to exhibit in vivo efficacy in several animal models of arthritis and hyperalgesia in rat and rabbit.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pelletier, J.P., et al. 2003. Arthritis Rheum.48, 1582.
Ciruela, A., et al. 2003. Br. J. Pharmacol.138, 751.
Bhattachar, S.N., et al. 2002. Int. J. Pharm.236, 135.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service